Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001045.hg.1 |
TC02000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001045.hg.1 |
ENST00000155926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001045.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001045.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001045.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001045.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001051.hg.1 |
TC02000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001051.hg.1 |
ENST00000155926.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001051.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001051.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001051.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02001051.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146359.hg.1 |
TC0200006757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200006757.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
ENST00000483034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146360.hg.1 |
TC0200006757.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
ENST00000155926.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
ENST00000381465.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
ENST00000405331.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
NM_021643.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
NR_027303.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200146367.hg.1 |
TC0200006757.hg.1 |
Clariom D TC |