Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143809.1 | NP_001137281.1 | 1 - 2 | 89 | 103 | |
GenBank mRNA | EF689012.1 | - | 1 - 2 | 89 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Homo sapiens Inventoried Probe spans exons |