Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015090.hg.1 |
ENST00000293845.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015090.hg.1 |
ENST00000539522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015090.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015090.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015090.hg.1 |
TC17001120.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000581598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000581598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
ENST00000581598.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015091.hg.1 |
ENST00000293845.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015091.hg.1 |
ENST00000539522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015091.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015091.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17015091.hg.1 |
TC17001120.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000581598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000581598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
ENST00000581598.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700009701.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175694.hg.1 |
TC1700009701.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
ENST00000293845.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
ENST00000539522.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
NM_001158261.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
NM_144681.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700175693.hg.1 |
TC1700009701.hg.1 |
Clariom D TC |