Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322177.1 | NP_001309106.1 | 2 | 5087 | 91 | |
NM_177966.6 | NP_808881.3 | 3 | 4914 | 91 | ||
GenBank mRNA | U79276.1 | - | 1 | 1049 | 91 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dynein axonemal heavy chain 12 | DNAH12 |
DHC3, DLP12, DNAH12L, DNAH7L, DNAHC12, DNAHC3, DNHD2, HDHC3, HL-19, HL19 |
-17.6kb | Anti-sense | Chr. 3 - 57293699 - 57545203 on NCBI Build GRCh38 |
Downstream | ADP ribosylation factor 4 | ARF4 | ARF2 | 7.8kb | Anti-sense | Chr. 3 - 57571363 - 57597488 on NCBI Build GRCh38 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |