Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transport and golgi organization 2 homolog | TANGO2 |
C22orf25, MECRCN |
Sense | Chr. 22 - 20017000 - 20067164 on NCBI Build GRCh38 | |
Downstream | microRNA 185 | MIR185 |
MIRN185, miR-185 |
8.2kb | Sense | Chr. 22 - 20033139 - 20033220 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |