Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEP domain containing MTOR-interacting protein | DEPTOR |
DEP.6, DEPDC6 |
-316.5kb | Sense | Chr. 8 - 119873655 - 120056202 on NCBI Build GRCh38 |
Downstream | mitochondrial ribosomal protein L13 | MRPL13 |
L13, L13A, L13mt, RPL13, RPML13 |
28.1kb | Anti-sense | Chr. 8 - 120395844 - 120445407 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |