Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
RAB10, member RAS oncogene family |
RAB10 | Sense | Chr. 2 - 26033860 - 26137454 on NCBI Build GRCh38 | ||
Downstream | GRB2 associated regulator of MAPK1 subtype 2 | GAREM2 |
FAM59B, GAREML |
95.1kb | Sense | Chr. 2 - 26173091 - 26202405 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |