Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | gamma-glutamylcyclotransferase | GGCT |
C7orf24, CRF21, GCTG, GGC |
-10.5kb | Anti-sense | Chr. 7 - 30496621 - 30504841 on NCBI Build GRCh38 |
Downstream | uncharacterized LOC401320 | LOC401320 | 32.5kb | Anti-sense | Chr. 7 - 30548357 - 30577779 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |