Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ST20-MTHFS readthrough | ST20-MTHFS | Sense | Chr. 15 - 79843547 - 79923106 on NCBI Build GRCh38 | ||
Downstream | ST20 antisense RNA 1 | ST20-AS1 | C15orf37 | 684.0 | Anti-sense | Chr. 15 - 79922771 - 79924854 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |