Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sideroflexin 2 | SFXN2 | -24.3kb | Sense | Chr. 10 - 102714518 - 102739189 on NCBI Build GRCh38 | |
Downstream | cytochrome P450 family 17 subfamily A member 1 | CYP17A1 |
CPT7, CYP17, P450C17, S17AH |
66.8kb | Anti-sense | Chr. 10 - 102830531 - 102837533 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |