Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kinesin family member 16B | KIF16B |
C20orf23, KISC20ORF, SNX23 |
-160.5kb | Anti-sense | Chr. 20 - 16272104 - 16573457 on NCBI Build GRCh38 |
Downstream | otoraplin | OTOR |
FDP, MIAL1 |
10.3kb | Sense | Chr. 20 - 16748353 - 16752164 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |