Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cytochrome b5 domain containing 1 | CYB5D1 | Anti-sense | Chr. 17 - 7857746 - 7864417 on NCBI Build GRCh38 | ||
Downstream | chromodomain helicase DNA binding protein 3 | CHD3 |
Mi-2a, Mi2-ALPHA, ZFH |
21.0kb | Anti-sense | Chr. 17 - 7884764 - 7912757 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |