Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK125525.1 | - | 1 | 1353 | 77 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
zinc finger CCCH-type, RNA binding motif and serine/arginine rich 2 |
ZRSR2 |
U2AF1-RS2, U2AF1L2, U2AF1RS2, URP, ZC3H22 |
-2.8kb | Anti-sense | Chr. X - 15789723 - 15825718 on NCBI Build GRCh38 |
Downstream | gastrin releasing peptide receptor | GRPR |
BB2, BB2R |
294.0kb | Anti-sense | Chr. X - 16123301 - 16153518 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |