Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | serine and arginine rich splicing factor 12 | SRSF12 |
SFRS13B, SFRS19, SRrp35 |
Anti-sense | Chr. 6 - 89095957 - 89118081 on NCBI Build GRCh38 | |
Downstream | gamma-aminobutyric acid type A receptor rho1 subunit | GABRR1 | 76.4kb | Anti-sense | Chr. 6 - 89177504 - 89231288 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |