Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | stabilin 2 | STAB2 |
FEEL2, FELE-2, FELL2, FEX2, HARE, SCARH1 |
-51.3kb | Anti-sense | Chr. 12 - 103587250 - 103766724 on NCBI Build GRCh38 |
Downstream |
tetratricopeptide repeat domain 41, pseudogene |
TTC41P |
GNN, GNNP |
25.3kb | Sense | Chr. 12 - 103843749 - 103930211 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |