Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019585.hg.1 |
TC09001561.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019585.hg.1 |
ENST00000304833.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09019585.hg.1 |
NM_001004450.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011444.hg.1 |
ENST00000304833.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011444.hg.1 |
ENST00000623530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011444.hg.1 |
ENST00000623530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011444.hg.1 |
ENST00000624439.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011444.hg.1 |
NM_001004450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011444.hg.1 |
ENST00000304833.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011444.hg.1 |
ENST00000623530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011444.hg.1 |
ENST00000623530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011444.hg.1 |
ENST00000624439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011444.hg.1 |
NM_001004450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
ENST00000304833.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
ENST00000623530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
ENST00000624439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
NM_001004450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
ENST00000304833.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
ENST00000623530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
ENST00000624439.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
NM_001004450.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900178011.hg.1 |
TC0900011444.hg.1 |
Clariom D TC |