Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033810.hg.1 |
TC03001870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033810.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033810.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033808.hg.1 |
TC03001870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033808.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033808.hg.1 |
ENST00000482567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033808.hg.1 |
ENST00000492200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033808.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033809.hg.1 |
TC03001870.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033809.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033809.hg.1 |
ENST00000482567.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033809.hg.1 |
ENST00000492200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033809.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195180.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195180.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195180.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195180.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195180.hg.1 |
TC0300012730.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012730.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195179.hg.1 |
TC0300012730.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
ENST00000443512.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
ENST00000482567.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
ENST00000492200.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
NM_001085420.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300195177.hg.1 |
TC0300012730.hg.1 |
Clariom D TC |