Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006273.hg.1 |
TC06000563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006273.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006273.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006274.hg.1 |
TC06000563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006274.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006274.hg.1 |
ENST00000426505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006274.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006272.hg.1 |
TC06000563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006272.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006272.hg.1 |
ENST00000426505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006272.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006268.hg.1 |
TC06000563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006268.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006269.hg.1 |
TC06000563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006269.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006271.hg.1 |
TC06000563.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006271.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006271.hg.1 |
ENST00000426505.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06006271.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155302.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155302.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155302.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155302.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155302.hg.1 |
TC0600007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600155303.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155303.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155303.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155303.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155303.hg.1 |
TC0600007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600155300.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155300.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155300.hg.1 |
TC0600007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600155301.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155301.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155301.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600155301.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155301.hg.1 |
TC0600007986.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
ENST00000244669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007986.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155299.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155299.hg.1 |
NM_006789.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600155299.hg.1 |
TC0600007986.hg.1 |
Clariom D TC |