Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006716519.3 | XP_006716582.1 | 2 - 3 | 6421 | 107 | |
XM_017013171.1 | XP_016868660.1 | 2 - 3 | 1155 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |