Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017093.hg.1 |
TC0X001265.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017093.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017093.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171399.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171399.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171399.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171399.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171399.hg.1 |
TC0X00010481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
ENST00000302917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010481.hg.1 |
NM_031273.2 |
Refseq |