Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002753.hg.1 |
TC01000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002753.hg.1 |
ENST00000361079.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002753.hg.1 |
NM_001012276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002757.hg.1 |
TC01000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002757.hg.1 |
ENST00000361079.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002757.hg.1 |
NM_001012276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002757.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002756.hg.1 |
TC01000166.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002756.hg.1 |
ENST00000361079.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002756.hg.1 |
NM_001012276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002756.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148172.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148172.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148172.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148172.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148172.hg.1 |
TC0100006912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148177.hg.1 |
TC0100006912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000357367.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000614831.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012946.hg.1 |
ENST00000621519.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100006912.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148175.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148175.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148175.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148175.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148175.hg.1 |
TC0100006912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
ENST00000616979.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
NM_001012277.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100148176.hg.1 |
TC0100006912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100148174.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148174.hg.1 |
ENST00000330881.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100148174.hg.1 |
TC0100006912.hg.1 |
Clariom D TC |