Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004617.3 | NP_004608.1 | 3 - 4 | 691 | 91 | |
GenBank mRNA | AK315464.1 | - | 3 - 4 | 578 | 91 | |
BC001386.2 | - | 3 - 4 | 561 | 3051146 | 91 | |
BT007430.1 | - | 3 - 4 | 394 | 91 | ||
U31449.1 | - | 3 - 4 | 559 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |