Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GZMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002104.2 | NP_002095.1 | 2 | 248 | 77 | |
GenBank mRNA | AK312074.1 | - | 2 | 271 | 77 | |
BC035802.1 | - | 2 | 248 | 5748080 | 77 | |
U26174.1 | - | 2 | 244 | 77 | ||
U35237.1 | - | 2 | 204 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons |