Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270952.1 | NP_001257881.1 | 4 - 5 | 488 | 102 | |
NM_006002.4 | NP_005993.1 | 4 - 5 | 410 | 102 | ||
XM_011535212.1 | XP_011533514.1 | 4 - 5 | 410 | 102 | ||
XM_011535213.2 | XP_011533515.1 | 3 - 4 | 391 | 102 | ||
XM_011535214.2 | XP_011533516.1 | 4 - 5 | 478 | 102 | ||
XM_017020725.1 | XP_016876214.1 | 4 - 5 | 410 | 102 | ||
XM_017020726.1 | XP_016876215.1 | 4 - 5 | 410 | 102 | ||
XM_017020727.1 | XP_016876216.1 | 3 - 4 | 385 | 102 | ||
GenBank mRNA | AK313665.1 | - | 4 - 5 | 400 | 102 | |
BC018125.2 | - | 4 - 5 | 369 | 3887201 | 102 | |
BQ427804.1 | - | 4 - 5 | 424 | 102 | ||
BT019359.1 | - | 4 - 5 | 339 | 102 | ||
CR456855.1 | - | 4 - 5 | 339 | 102 | ||
DA820150.1 | - | 4 - 5 | 410 | 102 | ||
M30496.1 | - | 4 - 5 | 359 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |