Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 7702 | MIR7702 | hsa-mir-7702 | Sense | Chr. 9 - 111271156 - 111271214 on NCBI Build GRCh38 | |
Downstream | olfactory receptor family 2 subfamily K member 2 | OR2K2 |
HSHTPCRH06, HTPCRH06, OR2AN1P, OR2AR1P |
54.4kb | Sense | Chr. 9 - 111326884 - 111330224 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |