Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000533564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000533564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000533564.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000310521.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000378966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000533564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000533564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000533564.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
ENST00000610924.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286341.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_001286342.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007074.hg.1 |
NM_152719.2 |
Refseq |