Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
ENST00000239878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
ENST00000379649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
ENST00000437952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286706.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_016617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NR_104584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NR_104585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
ENST00000239878.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
ENST00000379649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
ENST00000437952.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286703.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286704.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286705.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_001286706.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NM_016617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NR_104584.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
NR_104585.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300146974.hg.1 |
TC1300006914.hg.1 |
Clariom D TC |