Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104601.1 | 6 - 7 | 623 | 89 | ||
XM_011518515.2 | XP_011516817.1 | 11 - 12 | 1721 | 89 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PSMD5 antisense RNA 1 (head to head) | PSMD5-AS1 | -6.7kb | Anti-sense | Chr. 9 - 120843042 - 120854373 on NCBI Build GRCh38 | |
Downstream | TNF receptor associated factor 1 | TRAF1 |
EBI6, MGC:10353 |
39.6kb | Sense | Chr. 9 - 120902393 - 120929173 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |