Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187978.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187978.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187978.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187978.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187978.hg.1 |
TC1700010938.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187971.hg.1 |
TC1700010938.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700010938.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
ENST00000575039.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
ENST00000576345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
NR_110880.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
NR_110881.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700187972.hg.1 |
TC1700010938.hg.1 |
Clariom D TC |