Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120596.1 | 1 - 2 | 490 | 120 | ||
GenBank mRNA | AV709904.1 | - | 1 - 2 | 481 | 120 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transcription factor 20 | TCF20 |
AR1, SPBP, TCF-20 |
-80.5kb | Sense | Chr. 22 - 42160013 - 42283927 on NCBI Build GRCh38 |
Downstream | NFAT activating protein with ITAM motif 1 | NFAM1 | CNAIP | 11.2kb | Sense | Chr. 22 - 42380407 - 42438155 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons |