Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_121648.1 | 1 | 522 | 132 | ||
GenBank mRNA | BU658638.1 | - | 1 | 532 | 132 | |
KF110790.1 | - | 1 | 3199 | 132 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hemoglobin subunit beta pseudogene 1 | HBBP1 |
HBH1, HBHP |
-962.0 | Sense | Chr. 11 - 5241954 - 5243592 on NCBI Build GRCh38 |
Downstream | hemoglobin subunit gamma 1 | HBG1 |
HBG-T2, HBGA, HBGR, HSGGL1, PRO2979 |
2.7kb | Sense | Chr. 11 - 5248272 - 5249857 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |