Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_123733.1 | 10 - 11 | 738 | 97 | ||
NR_123734.1 | 9 - 10 | 708 | 97 | |||
GenBank mRNA | BU929788.1 | - | 9 - 10 | 512 | 97 | |
DN996275.1 | - | 10 - 11 | 560 | 97 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | activation-induced cytidine deaminase | AICDA |
AID, ARP2, CDA2, HEL-S-284, HIGM2 |
-33.1kb | Sense | Chr. 12 - 8602166 - 8612970 on NCBI Build GRCh38 |
Downstream | ribosomal modification protein rimK like family member B | RIMKLB |
FAM80B, NAAGS, NAAGS-I |
33.1kb | Anti-sense | Chr. 12 - 8681606 - 8783098 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |