Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000222.2 | NP_000213.1 | 4 | 718 | 104 | |
NM_001093772.1 | NP_001087241.1 | 4 | 718 | 104 | ||
XM_005265740.1 | XP_005265797.1 | 4 | 689 | 104 | ||
XM_005265741.1 | XP_005265798.1 | 4 | 689 | 104 | ||
XM_005265742.2 | XP_005265799.1 | 4 | 689 | 104 | ||
XM_017008178.1 | XP_016863667.1 | 4 | 689 | 104 | ||
XM_017008179.1 | XP_016863668.1 | 4 | 689 | 104 | ||
XM_017008180.1 | XP_016863669.1 | 4 | 689 | 104 | ||
GenBank mRNA | AK304031.1 | - | 4 | 689 | 104 | |
BC071593.1 | - | 4 | 707 | 4375615 | 104 | |
EU826594.1 | - | 4 | 631 | 104 | ||
GU983671.1 | - | 4 | 636 | 104 | ||
HM015525.1 | - | 4 | 636 | 104 | ||
HM015526.1 | - | 4 | 636 | 104 | ||
KT326922.1 | - | 4 | 649 | 104 | ||
X06182.1 | - | 4 | 652 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons |