Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53INP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568772.2 | XP_005568829.1 | 3 - 4 | 525 | 115 | |
XM_005568773.2 | XP_005568830.1 | 3 - 4 | 583 | 115 | ||
XM_005568774.2 | XP_005568831.1 | 3 - 4 | 703 | 115 | ||
XM_005568779.2 | XP_005568836.1 | 3 - 4 | 371 | 115 | ||
XM_005568780.2 | XP_005568837.1 | 2 - 3 | 413 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |