Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005560200.2 | XP_005560257.1 | 8 - 9 | 1445 | 82 | |
XM_005560201.2 | XP_005560258.1 | 7 - 8 | 1337 | 82 | ||
XM_015453158.1 | XP_015308644.1 | 6 - 7 | 1988 | 82 | ||
XM_015453159.1 | XP_015308645.1 | 8 - 9 | 1458 | 82 | ||
GenBank mRNA | AB169845.1 | - | 7 - 8 | 930 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |