Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005546036.2 | XP_005546093.1 | 1 - 2 | 1562 | 86 | |
XM_005546039.2 | XP_005546096.1 | 1 - 2 | 1562 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |