Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005569345.2 | XP_005569402.1 | 2 - 3 | 611 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |