Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568315.2 | XP_005568372.1 | 3 | 3693 | 155 | |
XM_005568316.2 | XP_005568373.1 | 2 | 3500 | 155 | ||
XM_015430090.1 | XP_015285576.1 | 3 | 4248 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |