Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287718.1 | NP_001274647.1 | 2 | 3156 | 81 | |
XM_005592970.2 | XP_005593027.1 | 3 | 4950 | 81 | ||
XM_015443655.1 | XP_015299141.1 | 2 | 4774 | 81 | ||
GenBank mRNA | AB445664.1 | - | 1 | 3058 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |