Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283438.1 | NP_001270367.1 | 4 - 5 | 1344 | 65 | |
GenBank mRNA | AF104339.1 | - | 4 - 5 | 1344 | 65 |
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350029.1 | NP_001336958.1 | 4 - 5 | 1310 | 65 |
Gene Symbol: UGT1A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350028.1 | NP_001336957.1 | 4 - 5 | 1400 | 65 | |
GenBank mRNA | AF104337.1 | - | 4 - 5 | 1401 | 65 |
Gene Symbol: UGT1A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350023.1 | NP_001336952.1 | 4 - 5 | 1323 | 65 | |
GenBank mRNA | AF104338.1 | - | 4 - 5 | 1324 | 65 |
Gene Symbol: UGT1A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001350026.1 | NP_001336955.1 | 4 - 5 | 1357 | 65 | |
GenBank mRNA | AF104336.1 | - | 4 - 5 | 1357 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |