Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102136135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005544397.2 | XP_005544454.1 | 5 - 6 | 1142 | 77 | |
XM_005544398.2 | XP_005544455.1 | 5 - 6 | 1134 | 77 | ||
XM_005544399.2 | XP_005544456.1 | 5 - 6 | 1128 | 77 | ||
XM_005544400.2 | XP_005544457.1 | 5 - 6 | 1133 | 77 | ||
XM_005544401.2 | XP_005544458.1 | 5 - 6 | 1137 | 77 | ||
XM_005544402.2 | XP_005544459.1 | 5 - 6 | 906 | 77 | ||
XM_005544405.2 | XP_005544462.1 | 5 - 6 | 894 | 77 | ||
XM_015440643.1 | XP_015296129.1 | 4 - 5 | 561 | 77 | ||
XM_015440666.1 | XP_015296152.1 | 5 - 6 | 1136 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |