Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102131468 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549700.2 | XP_005549757.1 | 4 - 5 | 1003 | 73 | |
XM_005549701.2 | XP_005549758.1 | 5 - 6 | 463 | 73 | ||
XM_005549703.2 | XP_005549760.1 | 5 - 6 | 1024 | 73 | ||
XM_015447484.1 | XP_015302970.1 | 7 - 8 | 1237 | 73 | ||
XM_015447485.1 | XP_015302971.1 | 5 - 6 | 307 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |