Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIPF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285289.1 | NP_001272218.1 | 5 - 6 | 724 | 73 | |
XM_005558118.2 | XP_005558175.1 | 5 - 6 | 1074 | 73 | ||
GenBank mRNA | AB169519.1 | - | 5 - 6 | 764 | 73 | |
AB171857.1 | - | 5 - 6 | 724 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |