Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592628.2 | XP_005592685.1 | 2 - 3 | 290 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |