Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589689.2 | XP_005589746.1 | 3 - 4 | 1032 | 99 | |
XM_005589691.1 | XP_005589748.1 | 3 - 4 | 565 | 99 | ||
XM_015441434.1 | XP_015296920.1 | 2 - 3 | 2484 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |