Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583464.2 | XP_005583521.1 | 5 | 2185 | 134 | |
XM_005583465.2 | XP_005583522.1 | 4 | 2053 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |