Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583558.2 | XP_005583615.1 | 2 - 3 | 2365 | 126 | |
XM_005583559.2 | XP_005583616.1 | 2 - 3 | 2397 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |