Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005541602.2 | XP_005541659.1 | 2 | 381 | 102 | |
XM_005541603.2 | XP_005541660.1 | 2 | 391 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |