Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005572134.2 | XP_005572191.1 | 1 | 1675 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |