Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005543380.2 | XP_005543437.1 | 1 - 2 | 224 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |